Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Edvotek™ Morphology of Cancer Cells
SDPSCI_ED

Catalog No. S67199 Shop All EDVOTEK Products
Change view
Click to view available options
Grade:
9 to Undergraduate
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S67199 9 to Undergraduate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S67199 Supplier Edvotek™ Supplier No. 990
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

  • All necessary materials plus complete instructions, background information, and study questions.

Requires

  • Microscope with 400–600X magnification
  • Forceps
  • Gloves
  • Distilled water
  • Differentiation of Human Cells by Retinoic Acid (S67198) also requires immersion oil and immersion objectives for the microscope

This experiment allows students to see the differences between normal and cancer cells in both their growth and cell types.

Provide an understanding and visual comparison of the morphology of various normal and transformed mammalian cells by microscopic examination. Procedures similar to these are used to examine biopsy specimens.

Analysis and Comparison of Various Mammalian Cell Types

  • Fixes and stains four different cell types for direct microscopic comparison to show the differences between normal and transformed cell cultures

Morphology of Cancer Cells

  • Compares the contact inhibition of normal cells to the increased growth of cancer cell colonies

Specifications

Grade 9 to Undergraduate
For Use With (Application) Students observe the differences between normal and cancer cells in both their growth and cell types
Includes Instructions, multispot slides (2 cell types each), fixing agent, eosin and flashblue™ stain, mounting medium, cover slips, transfer pipets, immersion troughs
Time Range 35 min.
Content And Storage Room temperature
Class Size 6 Lab Groups
Product Type Morphology of Cancer Cells
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.