Learn More
Teknova Mammalian Cell lysis Buffer, 100mL, Sterile
Supplier: Teknova 20741
500mM Tris Base,2.0M Sodium Chloride,5.0% Tween-20,Adjusted to pH 8.5,10X concentration,Volume provided is sufficient for 1 L of cell culture harvest
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.