Learn More
Diagnostic Biosystems MAMMOGLOBIN A Mammaglobin A
Supplier:  Diagnostic Biosystems RMAB03605
Encompass_Preferred
The mammaglobin gene was first identified using a differential screening approach directed at the isolation of novel, human breast cancer-associated genes. Mammaglobin-A is highly overexpressed in breast cancer cell lines and primary breast tumors. This pattern of expression is restricted to mammary epithelium and metastatic breast tumors. Thus, mammaglobin-Aspecific T cell immune responses may provide an important approach for the design of breast cancer-specific immunotherapy.
Catalog No. 50-238-5822
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.