Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MAN1B1 Monoclonal Antibody (3C9F10), Proteintech
SDP

Catalog No. 502332826 Shop All Proteintech Group Inc Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-233-2826 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-233-2826 Supplier Proteintech Group Inc Supplier No. 681211IG150UL
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MAN1B1 Monoclonal antibody specifically detects MAN1B1 in Human samples. It is validated for Western Blot

Specifications

Antigen MAN1B1
Applications Western Blot
Classification Monoclonal
Clone 3C9F10
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Accession No. Q9UKM7
Gene Alias ERMAN1; ERManI; MAN1B1; MANA-ER; MRT15; UNQ747/PRO1477
Host Species Mouse
Purification Method Protein A
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11253
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.