Learn More
Invitrogen™ MAN1B1 Polyclonal Antibody

Description
The antibody detects endogenous levels of total MAN1B1 protein.
Specifications
Specifications
| Antigen | MAN1B1 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | MAN1B1 |
| Gene Accession No. | A2AJ15, Q9UKM7 |
| Gene Alias | 3.2.1.113; E430019H13Rik; endoplasmic Reticulum Class I alpha-mannosidase; endoplasmic reticulum mannosyl-oligosaccharide 1,2-alpha-mannosidase; endoplasmic reticulum mannosyl-oligosaccharide 1,2-alpha-mannosidase 1; ER alpha 1,2-mannosidase; ER alpha-1,2-mannosidase; ER mannosidase 1; ERMan1; ERManI; Gm108; MAN1B1; Man9GlcNAc2-specific processing alpha-mannosidase; man9GlcNAc2-specific-processing alpha-mannosidase; MANA-ER; Mannosidase alpha class 1B member 1; mannosidase, alpha, class 1B, member 1; MRT15; RGD1563595; RP11-350O14.2; UNQ747/PRO1477 |
| Gene Symbols | MAN1B1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.