Learn More
Invitrogen™ MAN2A1 Polyclonal Antibody

Description
Antibody detects endogenous levels of total Mannosidase II.
Specifications
Specifications
| Antigen | MAN2A1 |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide |
| Gene | MAN2A1 |
| Gene Accession No. | P27046, P28494, Q16706 |
| Gene Alias | Alpha-mannosidase 2; alpha-mannosidase II; AMan II; Golgi alpha-mannosidase II; golgi integral membrane protein 7; GOLIM7; man II; Man2a1; Mana2; Mana-2; MANII; Manno; mannosidase 2, alpha 1; mannosidase alpha 2; mannosidase alpha class 2A member 1; mannosidase, alpha type II; mannosidase, alpha, class 2A, member 1; mannosyl-oligosaccharide 1,3-1,6-alpha-mannosidase; Map-2 |
| Gene Symbols | MAN2A1 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.