Learn More
TARGETMOL CHEMICALS INC MANAGLINAT DIALANETIL 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T16009L
Encompass_Preferred
Also available in 1mg 10mg 25mg 50mg 100mg 500mg and bulk. Please contact Fisher for quotes. Managlinat dialanetil (MB06322) is an orally available and potent fructose 16-bisphosphatase (FBPase) inhibitor with hypoglycaemic activity for the study of type 2 diabetes. purity: 98%
Catalog No. 50-257-7347
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.