Learn More
AdipoGen Mancozeb
SDP
Supplier:  AdipoGen CDXM0158M500
Encompass_Preferred
Chemical. CAS 8018-01-7. Formula C4H6MnN2S4 . C4H6N2S4Zn. MW 265.30 . 275.75. Synthetic. Mancozeb is a dithiocarbamate non-systemic agricultural fungicide with multi-site, protective action on contact. It is a combination of two dithiocarbamates maneb and zineb. The mixture controls many fungal diseases in a wide range of field crops, fruits, nuts, vegetables and ornamentals. It reacts with, and inactivates, the sulfhydryl groups of amino acids and enzymes of fungal cell, resulting in disruption of lipid metabolism, respiration and production of adenosine triphosphate. Shown to by cytotoxic, induce apoptosis and damaging DNA on mammalian cells. also shown to have neurotoxic and genotoxic activity.
Catalog No. 50-256-0486
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.