Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Mandelic acid (2-hydroxy-2-phenylacetic acid) | 90-64-2 | MFCD00064250 | 99.9% | 152.15 g/mol | C8H8O3 | 1 ML
SDP

Supplier:  Medchemexpress LLC HYW01559110MM/1ML

Encompass_Preferred

Mandelic acid (2-hydroxy-2-phenylacetic acid) is an alpha-hydroxycarboxylic acid used in antimicrobial research and as an intermediate in pharmaceutical and fine-chemical synthesis. Supplied as a ready-to-use 10 mM solution (1 mL) in DMSO, it provides a consistent reagent for in vitro studies and formulation work.

  • Ready-to-use 10 mM solution in DMSO, 1 mL volume.
  • High purity (99.92%) for reliable experimental performance.
  • CAS number 90-64-2 for clear substance identification.
  • Chemical formula C8H8O3 and molecular weight 152.15 g/mol.
  • Suitable for antimicrobial research and intermediate synthesis.
  • Soluble in DMSO and compatible with common in vitro co-solvents.

Catalog No. 50-003-00665


May include imposed supplier surcharges.
Only null left
Add to Cart

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.