Learn More
Medchemexpress LLC Mandelic acid-2,3,4,5,6-d5 | 70838-71-0 | 98.4% | 157.18 | C8H3D5O3 | 100 MG
SDP
Supplier:  Medchemexpress LLC HYW015591S100MG
Encompass_Preferred
Mandelic acid-2,3,4,5,6-d5 is a deuterium-labeled (d5) analogue of mandelic acid used as a stable isotope internal standard and tracer in analytical chemistry and mass spectrometry. It is supplied with high purity and characterized molecular properties for use in quantitative calibration, method development, and metabolic or pharmacokinetic studies.
  • Deuterium-labeled analogue for use as an internal standard.
  • High purity suitable for quantitative assays.
  • Consistent isotopic labeling for accurate mass spectrometry analysis.
  • Appropriate package sizes for method development and reference use.
  • Characterized molecular properties to support analytical workflows.
Catalog No. 50-003-00402
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.