Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Mandelic acid-2,3,4,5,6-d5 | 70838-71-0 | 98.4% | 157.18 | C8H3D5O3 | 10 MG
SDP

Supplier:  Medchemexpress LLC HYW015591S10MG

Encompass_Preferred

Mandelic acid-2,3,4,5,6-d5 is a deuterium-labeled analogue of mandelic acid used as an internal standard and tracer in analytical workflows such as mass spectrometry. Supplied as a white to off-white solid, it provides a stable isotopic reference for quantitative assays and method development.

  • Used as internal standard for mass spectrometry and isotopic tracing.
  • High chemical purity (98.4%).
  • White to off-white solid for convenient handling and storage.
  • Molecular formula C8H3D5O3 and molecular weight 157.18.
  • Powder stable at -20°C for up to 3 years, with defined solvent storage conditions.

Catalog No. 50-003-00730


May include imposed supplier surcharges.
Only null left
Add to Cart

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.