Learn More
Medchemexpress LLC Mandelic acid | 90-64-2 | MFCD00064250 | 99.9% | 152.15 g/mol | C8H8O3 | 50 G
Supplier: Medchemexpress LLC HYW01559150G
Mandelic acid (CAS 90-64-2) is an alpha-hydroxycarboxylic acid used as a research reagent and synthetic intermediate in pharmaceutical and fine-chemical applications. It shows antimicrobial activity and is supplied with batch-specific quality documentation for analytical and formulation work.
- High purity (99.92% assay by HPLC).
- White crystalline solid, suitable for analytical use.
- Commonly used as a synthetic intermediate and research reagent.
- Soluble in polar organic solvents, facilitating formulations.
- Provided with batch-specific certificate of analysis for QC.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.