Learn More
Strem, An Ascensus Company CAS 793718-16-8. 2g. (SS)-(-)-22-Bis[(R)-(NN-dimethylamino)(phenyl)methyl]-11-bis(di(35-dimethylphenyl)phosphino)ferrocene min. 97%. MFCD07369030
SDP
Supplier:  Strem, An Ascensus Company 2602462G
Encompass_Preferred
CAS 793718-16-8. 2g. (SS)-(-)-22-Bis[(R)-(NN-dimethylamino)(phenyl)methyl]-11-bis(di(35-dimethylphenyl)phosphino)ferrocene min. 97%. MFCD07369030. Molecular Weight 932.99. Molecular Formula C60H66FeN2P2. Color/form orange pwdr. Strem 26-0246. http//www.strem.com/catalog/v/26-0246/
Catalog No. 50-211-0788
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.