Learn More
SPEX CERTIPREP LLC 3P MANGANESE 10000UG/ML STD 100MG
Supplier:  SPEX CERTIPREP LLC 3P 10M321100
Encompass_Preferred
(HPS by Zeptometrix) Mn 10Mppm in 4% HNO3 Exp Date: 18 Months This standard is traceable to NIST and certified Guide 34 and 17025.
Catalog No. NC0664533
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.