Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Manganese, 99.5%, MP Biomedicals

Catalog No. ICN211997 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN211997 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN211997 Supplier MP Biomedicals, Inc Supplier No. 0521199780

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 7439-96-5
Molecular Formula Mn
Molecular Weight (g/mol) 54.94
MDL Number MFCD00011111
InChI Key PWHULOQIROXLJO-UHFFFAOYSA-N
Synonym colloidal, mangan, manganese, elemental, cutaval, magnacat, metal alloy, tronamang, fume, mangan polish, compounds
PubChem CID 23930
ChEBI CHEBI:35154
IUPAC Name manganese
SMILES [Mn]

Specifications

Quantity 100 g
Chemical Name or Material Manganese, 99.5%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.