Learn More
R&D Systems™ Manganese cation (2 x 50 ul)
SDP
Catalog No. 48109014
Change view
Click to view available options
Quantity:
3 x 30 μL
Catalog No. Quantity
48109014 3 x 30 μL
1 options
This item is not returnable. View return policy
Catalog No. 48109014 Supplier R&D Systems™ Supplier No. 48109014
Customers require a valid business account associated to their profile to access pricing and ordering options.
This item is not returnable. View return policy
Apoptosis Detection
3 vials of 30μL
Specifications
Quantity 3 x 30 μL
Product Type Manganese cation
Content And Storage Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.