Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Manganese Gluconate, Dihydrate, USP, 98-102%, Spectrum™ Chemical
SDP

Catalog No. 18604191 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-604-191 12 kg Poly Pail
18-604-189 500 g Poly Bottle
18-604-190 2.5 kg Poly Pail
18-604-192 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-604-191 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. M140412KG
Add to Cart
Add to Cart

M1404, 6485-39-8, C12H22MnO14·2H2O

Spectrum™ Chemical Manganese Gluconate, Dihydrate, USP - All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 6485-39-8

Specifications

CAS Max % 100%
Quantity 12 kg
Percent Purity 98 to 102%
Grade USP
Packaging Poly Pail
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.