Learn More
Manganese(II) iodide, anhydrous, 98%
Description
Manganese(II) iodide can be used as a pink pigment or as a source of the manganese ion or iodide ion. It is often used in the lighting industry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 638°C |
| Color | Red to White |
| Physical Form | Powder |
| Quantity | 10 g |
| Linear Formula | MnI2 |
| Merck Index | 14,5732 |
| Solubility Information | Soluble in water with gradual decomposition; Soluble in alcohol |
| Formula Weight | 308.75 |
| Percent Purity | 98% |
| Odor | Odorless |
| Show More |
Safety and Handling
| Hazard Category | Warning |
| Hazard Statement | GHS H Statement H302 Harmful if swallowed. |
| Precautionary Statement | GHS P Statement P264-P270-P301+P312-P330-P501a Wash thoroughly after handling. Do not eat, drink or smoke when using this product. IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. Rinse mouth. Dispose of contents/container in accordance with local/regional/national/international regulations. |
| EINECSNumber | 232-201-6 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.