Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Manganese(II) iodide, ultra dry, 99.99% (metals basis)

Catalog No. p-7052300
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4716603 1 g
AA4716606 5 g
AA4716609 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA4716603 Supplier Thermo Scientific Chemicals Supplier No. 04716603
Only null left
Add to Cart

CAS: 7790-33-2

It is often used in the lighting industry and it is a source of its derivatives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7790-33-2
MDL Number MFCD00016225

Specifications

Melting Point 638°C
Color Red to White
Physical Form Beads
Quantity 1 g
Merck Index 14,5732
Solubility Information Soluble in water.
Formula Weight 308.75
Percent Purity 99.99%
Odor Odorless
Packaging Ampule
Sensitivity Air, light, and moisture sensitive
Density 5.0 g/mL
Assay (metals basis)
Chemical Name or Material Manganese(II) iodide, Ultra dry
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement
H302
Harmful if swallowed.
Precautionary Statement GHS P Statement
P264-P270-P301+P312-P330-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 232-201-6
TSCA Yes
storageNote1
  • Ampouled under argon
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.