Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Manganese(II) iodide, ultra dry, 99.99% (metals basis)

Catalog No. AA3571004
Change view
Click to view available options
Quantity:
2 g
10 g
Packaging:
Ampouled under argon
Plastic bottle; Ampouled under argon
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AA3571004 2 g Ampouled under argon
AA3571009 10 g Plastic bottle; Ampouled under argon
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA3571004 Supplier Thermo Scientific Chemicals Supplier No. 03571004
Add to Cart

Often used in the lighting industry and it is a source of its derivatives. | CAS: 7790-33-2 | I2Mn

It is often used in the lighting industry and it is a source of its derivatives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7790-33-2
Molecular Formula I2Mn
MDL Number MFCD00016225

Specifications

Melting Point 638°C
Physical Form Powder or Flakes
Quantity 2 g
Linear Formula MnI2
Solubility Information Soluble in water.
Formula Weight 308.75
Percent Purity 99.99%
Packaging Ampouled under argon
Sensitivity Air, light, and moisture sensitive
Specific Gravity 5
Density 5
Chemical Name or Material Manganese(II) iodide
Show More Show Less
EINECSNumber 232-201-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.