Learn More
Manganese(II) sulfate monohydrate, 99%
Description
Manganese(II) sulfate monohydrate acts as a colorant in dyes. It is used in the preparation of manganese and manganese dioxide. It finds application for remediation of manganese-deficient soil. It is also used in dry-cell batteries.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Manganese(II) sulfate monohydrate acts as a colorant in dyes. It is used in the preparation of manganese and manganese dioxide. It finds application for remediation of manganese-deficient soil. It is also used in dry-cell batteries.
Solubility
Soluble in water and alcohol. Insoluble in ether.
Notes
Loses all water at 400-450°. Hygroscopic. Incompatible with strong oxidizing agents.
Specifications
Specifications
| pH | 3 to 3.5 |
| Quantity | 2500 g |
| UN Number | UN3077 |
| Merck Index | 14,5739 |
| Solubility Information | Soluble in water and alcohol. Insoluble in ether. |
| Formula Weight | 169.02 (151.00 Anhydrous) |
| Percent Purity | 99% |
| Odor | Odorless |
| Sensitivity | Hygroscopic |
| Density | 2.95 |
| Show More |
Safety and Handling
| Hazard Category | H318-H373 |
| Hazard Statement | GHS H Statement H373 May cause damage to organs through prolonged or repeated exposure. |
| Precautionary Statement | P260-P280-P305+P351+P338-P310-P314-P501c |
| DOTInformation | Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S. |
| EINECSNumber | 232-089-9 |
| RTECSNumber | OP0893500 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.