Learn More
Strem, An Ascensus Company CAS# 1336-93-2. 100g. Manganese naphthenate in mineral spirits (6% Mn). MFCD00147533
SDP
Supplier:  Strem, An Ascensus Company 932516100G
Encompass_Preferred
CAS# 1336-93-2. 100g. Manganese naphthenate in mineral spirits (6% Mn). MFCD00147533. Molecular Weight: . Molecular Formula: . Color/form: viscous liq. Strem# 93-2516. http://www.strem.com/catalog/v/93-2516/
Catalog No. 50-901-14600
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.