Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Manganese, plasma standard solution, Specpure™ Mn 1000μg/mL

Catalog No. p-4408567
Change view
Click to view available options
Quantity:
50 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA13826AD 50 mL
AA13826AE 100 mL
AA13826AP 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA13826AD Supplier Thermo Scientific Chemicals Supplier No. 013826AD
Only null left
Add to Cart

CAS: 7439-96-5 | Mn | 54.94 g/mol

Manganese, plasma standard solution, Specpure™, Mn 1000μg/ml is used as a standard solution in analytical chemistry and atomic absorption spectroscopy. It is also used as a single-element standard solution for plasma emission spectrometry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7439-96-5 , 7697-37-2 , 7732-18-5
Molecular Formula Mn
Molecular Weight (g/mol) 54.94
MDL Number MFCD00011111
InChI Key PWHULOQIROXLJO-UHFFFAOYSA-N
PubChem CID 23930
ChEBI CHEBI:35154
IUPAC Name manganese
SMILES [Mn]

Specifications

Vapor Pressure 23 hPa (17mm Hg) at 20°C
Physical Form Liquid
Color Colorless
Solubility Miscible with water
Quantity 50 mL
Concentration or Composition (by Analyte or Components) Nitric acid: 5.0%; Manganese: 0.1%; Water: 94.9%
Chemical Name or Material Manganese
Hazard Category H290-H315-H318
Precautionary Statement P234-P264b-P280-P302+P352-P305+P351+P338-P310-P332+P313-P362-P390
DOTInformation Hazard Class: 8, Packing Group: III
EINECSNumber 231-714-2
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.