Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Manganese Standard, 1000mg/L in 2% HNO3 (Assurance Grade), SPEX CertiPrep™
SDP

Catalog No. p-218948 Shop All Spex CertiPrep Products
Change view
Click to view available options
Quantity:
30 mL
250 mL
Packaging:
Poly Bottle
Triple-leached LDPE bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
PLMN22M 30 mL Triple-leached LDPE bottle
PLMN22T 250 mL Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PLMN22M Supplier SPEX CertiPrep Supplier No. PLMN22M
Only null left
Add to Cart
Add to Cart

Includes

NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
Mn1000μg/mL

Certifications

ISO/IEC 17025:2005, ISO 17034:2016, ISO 9001: 2015

Compliance

GHS/OSHA Complaint

Chemical Identifiers

CAS 7439-96-5 , 7697-37-2 , 7732-18-5
Molecular Formula Mn
Molecular Weight (g/mol) 54.94
MDL Number MFCD00011111
InChI Key PWHULOQIROXLJO-UHFFFAOYSA-N
PubChem CID 23930
ChEBI CHEBI:35154
IUPAC Name manganese
SMILES [Mn]

Specifications

Packaging Triple-leached LDPE bottle
Concentration or Composition Notes Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
CAS Min % 0.1, 2.0, 97.9
CAS Max % 0.1, 2.0, 97.9
Technique AA, ICP
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Type Certified Reference Material
Solvent or Matrix 2% HNO3
Quantity 30 mL
Grade Assurance
Chemical Name or Material Manganese Standard
Show More Show Less
Hazard Category Warning: Causes skin irritation, serious eye irritation Wear protective gloves/eye/face protection. If in eyes, rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. If skin/eye irritation occurs, get medical advice/attention.
DOTInformation Nonhazardous
Recommended Storage Ambient
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.