Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Manganese Sulfate Monohydrate, ACS Reagent (Manganous Sulfate), Reagents

Catalog No. 01337904
Change view
Click to view available options
Quantity:
100 g
500 g
3 kg
12 kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-337-904 500 g
01-337-907 100 g
01-337-906 12 kg
01-337-905 3 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 01-337-904 Supplier Reagents Holdings Llc Supplier No. C1243000500A5
Add to Cart
Add to Cart

Manganese Sulfate Monohydrate, ACS Reagent (Manganous Sulfate), ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 10034-96-5
Molecular Formula H2MnO5S
Molecular Weight (g/mol) 169.01
InChI Key ISPYRSDWRDQNSW-UHFFFAOYSA-L
Synonym Manganous Sulfate Monohydrate
IUPAC Name manganese(2+) hydrate sulfate
SMILES O.[Mn++].[O-]S([O-])(=O)=O

Specifications

Boiling Point 850°C
Melting Point 700°C
Color Pink
Physical Form Solid
Quantity 500 g
Grade ACS
Packaging Poly Bottle
Specific Gravity 2.95
Chemical Name or Material Manganese Sulfate Monohydrate, ACS Reagent (Manganous Sulfate)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.