Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Manganese Sulfate, Monohydrate, Reagent, ACS, 98-101%, Spectrum™ Chemical
SDP

Catalog No. 18604216 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
125 g
500 g
2.5 kg
6 x 500 g
12 kg
4 x 2.5 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-604-216 6 x 500 g Poly Bottle
18-604-214 125 g Poly Bottle
18-604-215 500 g Poly Bottle
18-604-217 2.5 kg Poly Pail
18-604-218 4 x 2.5 kg Poly Pail
18-604-219 12 kg Poly Pail
18-604-220 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 18-604-216 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. M1115500GMCS6
Add to Cart
Add to Cart

M1115, 10034-96-5, MnSO4·H2O

Spectrum™ Chemical Manganese Sulfate, Monohydrate, Reagent, ACS , is an inorganic compound that like many metal sulfates can form a variety of hydrates such as heptahydrates, monohydrates, tetrahydrates, pentahydrates. When these salts are dissolved, they tend to give a light pink solution, a characteristic of manganese sulfate. As an ACS grade Reagent, Spectrum Chemical manufactured manganese sulfate is used as the quality standard against which other substances are graded and has met the toughest regulatory standards for quality and pureness.

Chemical Identifiers

CAS 10034-96-5
Molecular Formula H2MnO5S
Molecular Weight (g/mol) 169.01
MDL Number MFCD00149159
InChI Key ISPYRSDWRDQNSW-UHFFFAOYSA-L
IUPAC Name manganese(2+) hydrate sulfate
SMILES O.[Mn++].[O-]S([O-])(=O)=O

Specifications

CAS Max % 100%
Quantity 6 x 500 g
Percent Purity 98 to 101%
Loss on Ignition 10.0-12%
Packaging Poly Bottle
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.