Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Manganous Sulfate, 0.31% (w/v), Ricca Chemical
SDP

Catalog No. 461016 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
500 mL
1 L
Packaging:
Natural Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
4610-16 500 mL Natural Poly Bottle
R46100001A 1 L Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 4610-16 Supplier Ricca Chemical Company Supplier No. 461016
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7732-18-5 , 7664-93-9 , 10034-96-5
Molecular Weight (g/mol) 318.68 g/mol
Synonym dihydrogen oxide, dihydrogen monoxide

Specifications

Color Colorless
CAS Min % 97.0, 0.54, 0.3
CAS Max % 100.0, 0.56, 0.31
Percent Purity 0.31%
Grade Laboratory
Packaging Natural Poly Bottle
Solvent or Matrix Aqueous Solution
Quantity 500 mL
Physical Form Liquid
Concentration or Composition Notes 0.31% (w/v)
Chemical Name or Material Manganous Sulfate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.