Learn More
Apexbio Technology LLC Manidipine,25mg CAS# 89226-50-6
SDP
Supplier:  Apexbio Technology LLC B141825
Encompass_Preferred
Manidipine is a calcium channel blocker that is used clinically as an antihypertensive.Manidipine is a dihydropyridine calcium antagonist, which causes systemic vasodilation by inhibiting the voltage-dependent calcium inward currents in smooth muscle cell. Other sizes are also available. Please inqury us for quote.
Catalog No. 50-224-0586
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.