Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Mannitol Salt Agar (Dehydrated)

Catalog No. OXCM085B Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
OXCM085B 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. OXCM085B Supplier Thermo Scientific™ Supplier No. CM0085B
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Presumptively detect, isolate and enumerate pathogenic staphylococci in milk, food, fish and other specimens with this highly selective medium.

Presumptively detect, isolate and enumerate pathogenic staphylococci in milk, food, fish and other specimens with Thermo Scientific™ Oxoid™ Mannitol Salt Agar (Dehydrated). Due to high concentrations of salt in the medium, most unwanted bacteria are inhibited with the exception of some halophilic marine organisms. Presumptive coagulase-positive staphylococci produce colonies surrounded by bright yellow zones while non-pathogenic staphylococci produce colonies with reddish purple zones.

Staphylococcus aureus is a coagulase-positive, catalase-positive, Gram-positive cocci that occur singly or in pairs, tetrads, short chains, and grape-like clusters. S. aureus can produce a variety of syndromes with clinical manifestations, including skin and soft tissue infections, empyema, bloodstream infections, pneumonia, osteomyelitis, septic arthritis, endocarditis, sepsis, and meningitis.

Clearly and easily differentiate Staphylococcus aureus and Staphylococcus epidermidis while most other bacteria is inhibited by the high salt concentration.

  • Highly selective: due to high concentrations of salt in the medium.
  • Easy to differentiate: Staphylococcus aureus forms yellow colonies with yellow halo while Staphylococcus epidermis forms pink colonies with medium turning pink.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand

TRUSTED_SUSTAINABILITY

Specifications

Description Mannitol Salt Agar
Form Powder
Product Type Mannitol Salt Agar
Quantity 500 g
Yield For 4.5L medium

For Laboratory Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.