Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Manual Crimpers, Decrimpers and Decrimping Pliers

Catalog No. 0337524AC
Change view
Click to view available options
Size:
8 mm
11 mm
13 mm
20 mm
28 mm
30 mm
32 mm
Type:
Manual Crimper
Manual Crimper (Flip-off)
Manual Decapping Plier
Manual Decrimper
26 product options available for selection
Product selection table with 26 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Type
03-375-24AC 20 mm Manual Decrimper
03-375-75C 13 mm Manual Crimper (Flip-off)
03-375-75D 20 mm Manual Crimper (Flip-off)
03-375-75B 32 mm Manual Crimper
06-406-19AD 8 mm Manual Decrimper
03-377-125 32 mm Manual Decrimper
13-622-409 8 mm Manual Crimper
13-622-407 11 mm Manual Crimper
13-622-408 20 mm Manual Crimper
06-406-19R 8 mm Manual Crimper
03-375-7 11 mm Manual Crimper
03-375-17A 13 mm Manual Crimper
03-375-24A 20 mm Manual Crimper
03-375-75A 28 mm Manual Crimper
03-375-52A 30 mm Manual Crimper
13-622-365 11 mm Manual Crimper
13-622-366 20 mm Manual Crimper
13-622-411 20 mm -
03-375-8A 11 mm Manual Decrimper
03-375-17D 13 mm Manual Decrimper
03-375-75F 28 mm Manual Decrimper
03-375-52C 30 mm Manual Decrimper
03-251-701 20 mm Manual Crimper
03-375-8 11 mm Manual Decapping Plier
03-375-17B 13 mm Manual Decapping Plier
03-375-24B 20 mm Manual Decapping Plier
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 26 options available.
26 missing translation for 'options'
Catalog No. 03-375-24AC missing translation for 'mfr' Thermo Scientific™ missing translation for 'supplierNo' C4020102

Achieve secure, repeatable crimps or remove seals with minimized risk of damage.

Achieve secure, repeatable crimps or remove seals with minimized risk of damage when you use Thermo Scientific™ Manual Crimpers, Decrimpers and Decrimping Pliers, also for use with National or Chromacol vials.

Manual Crimpers
Lightweight, precision crimpers attach seals to chromatography or serum vials. Available for standard aluminum or flip-off seals.
  • Factory set to properly crimp standard-thickness seals
  • Adjustable handle setting screw allows proper crimps with septa of varying thickness
Manual Decripmers
Forged pliers feature a fitted gripping surface designed to allow removal of the listed sizes of seals.
  • Simply grip pliers around top of seal, squeeze and pry upward to remove
Manual Decrimping Pliers
Place decrimping head squarely on seal and squeeze.
  • Minimizes damage to vial rim with chromatography or serum vials
  • Removes standard aluminum seals and many steel seals
TRUSTED_SUSTAINABILITY

Specifications

Description Manual Decrimper
No. per Pack 1 Ea.
Type Manual Decrimper
For Use With (Equipment) 20 mm Standard Seal
Size 20 mm
For Use With (Application) Industrial Chromatography
Product Line National

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.