Learn More
Thermo Scientific™ Manual Crimpers and Decrimpers for Sealing and Unsealing Crimp Vials
GSA_VA
Catalog No. 03452418
Change view
Click to view available options
For Use With (Equipment):
11 mm Crimp Vials
13 mm Crimp Vials
20 mm Crimp Vials
8 mm Crimp Vials
Type:
Manual Crimper
Manual Decrimper
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment) Type
03-452-418 11 mm Crimp Vials Manual Decrimper
03-452-414 11 mm Crimp Vials Manual Crimper
03-452-415 13 mm Crimp Vials Manual Crimper
03-452-416 20 mm Crimp Vials Manual Crimper
03-452-417 8 mm Crimp Vials Manual Crimper
03-452-419 13 mm Crimp Vials Manual Decrimper
03-452-420 20 mm Crimp Vials Manual Decrimper
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 03-452-418 Supplier Thermo Scientific™ Supplier No. 60180DCR11
Add to Cart
Add to Cart
Use Thermo Scientific™ Manual Vial Crimpers and Decrimpers to attach or remove crimp caps from vials, respectively.
Use Thermo Scientific™ Vial Crimpers and Decrimpers to manually attach or remove crimp caps from vials, respectively. These manual crimpers and decappers are available in 8 mm, 11 mm, 13 mm, and 20 mm sizes and allow you to easily seal or remove crimp caps reliably.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
TRUSTED_SUSTAINABILITY
Specifications
Type Manual Decrimper
For Use With (Equipment) 11 mm Crimp Vials
Material Stainless Steel/Aluminum
Product Line SureSTART Level 2