Learn More
AdipoGen Manumycin A
SDP
Supplier:  AdipoGen BVT0091M005
Encompass_Preferred
Chemical. CAS 52665-74-4. Formula C31H38N2O7. MW 550.6. Isolated from Streptomyces parvulus. Antibiotic. Apoptosis and endoplasmic reticulum stress-mediated cell death inducer. Potent, selective and competitive cell permeable rasfarnesyltransferase inhibitor IC50 = 30 nM. Does not affect geranylgeranyltransferase IC50 = 180 µM. Inhibition is competitive with respect to farnesyl pyrophosphate and non-competitive with respect to Ras. Neutral sphingomyelinase inhibitor. Blocks insulin-induced MAP kinase activation in rat cardiac myocytes 19 µM. Targets protein phosphatase 1alpha PP1alpha and reduces hydrogen peroxide. Corrects aberrant splicing of the muscle chloride channel Clcn1 in myotonic dystrophy type 1 DM1. Shows potential anti-inflammatory activity.
Catalog No. 50-256-2108
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.