Learn More
AdipoGen Manumycin A,Chemical. CAS: 52665-74-4
Supplier: AdipoGen BVT0091M001
Manumycin A, AdipoGen Life Sciences. Chemical. CAS: 52665-74-4. Formula: C31H38N2O7. MW: 550.6. Isolated from Streptomyces parvulus. Antibiotic. Apoptosis and endoplasmic reticulum stress-mediated cell death inducer. Potent, selective and competitive cell permeable rasfarnesyltransferase inhibitor (IC50 = 30 nM). Does not affect geranylgeranyltransferase (IC50 = 180 ?M). Inhibition is competitive with respect to farnesyl pyrophosphate and non-competitive with respect to Ras. Neutral sphingomyelinase inhibitor. Blocks insulin-induced MAP kinase activation in rat cardiac myocytes (19 ?M). Targets protein phosphatase 1alpha (PP1alpha) and reduces hydrogen peroxide. Corrects aberrant splicing of the muscle chloride channel Clcn1 in myotonic dystrophy type 1 (DM1). Shows potential anti-inflammatory activity.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.