Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Manumycin A, MP Biomedicals™

Catalog No. MP218366605 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP218366605 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP218366605 Supplier MP Biomedicals, Inc Supplier No. 0218366605
Add to Cart
Add to Cart

An antibiotic that act as a potent and selective farnesyltransferase inhibitor

  • By adding a farnesyl group to the -SH of the cysteine near the end of target proteins, Farnesyltransferase posttranslationally-modifies proteins to form a thioether linkages
  • Used as an antibiotic and different research branches
  • Farnesylation enables the proteins to become membrane associated
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 52665-74-4
Molecular Formula C31H38N2O7
Synonym Manumycin A, manumycin, Ucfi-C, AC1O5PHE, UNII-OIQ298X4XD

Specifications

Quantity 5 mg
Color Brown-Orange
Purity >95
Molecular Weight (g/mol) 550.65
Chemical Name or Material Manumycin A
Recommended Storage 4°C
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.