Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Manumycin A, Ras Farnesyltransferase Inhibitor, Solid, 5 mg

Catalog No. 502705908
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-270-5908 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-270-5908 Supplier Abcam Supplier No. AB1442975MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Potent, selective, and cell-permeable Ras farnesyltransferase inhibitor that induces apoptosis and exhibits antitumor effects in vivo.

  • Potent, selective, competitive cell-permeable Ras farnesyltransferase inhibitor (Ki = 1.2 μM).
  • IκB kinase inhibitor.
  • Antibiotic agent.
  • Induces apoptosis.
  • Shows antitumor effects in vivo.
  • Molecular weight: 550.6 Da.
  • Purity: >96%.
  • Physical form: Solid.

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Form Solid
Content And Storage Store at -20°C.
Shipping Condition Blue Ice
Substrate Properties Chemical Substrate
Target Molecule Ras Farnesyltransferase, IκB Kinase
Quantity 5 mg
For Use With (Application) Ras farnesyltransferase inhibition, IκB kinase inhibition, apoptosis induction, antitumor research, antibiotic research
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.