Learn More
Description
Western blot analysis in human brain using this antibody detected two main bands between 100-130 kDa corresponding to heavy and light chains of MAP1S.
Specifications
Specifications
| Antigen | MAP1S |
| Applications | Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
| Classification | Polyclonal |
| Concentration | 0.16 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | MAP1S |
| Gene Accession No. | Q66K74 |
| Gene Alias | BPY2 interacting protein 1, BPY2IP1, C19orf5, MAP 1S, MAP1S, MAP8, VCY2 interacting protein 1, VCY2IP 1, VCY2IP1 |
| Gene Symbols | MAP1S |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
