Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MAP2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. 01670262 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-670-262 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-670-262 Supplier Invitrogen™ Supplier No. PA110005
  • $706.50
    $547.65 / Each of 1

    Save $158.85

    22% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Chicken Polyclonal Antibody

PA1-10005 was raised against recombinant constructs of the entire human projection domain, and so recognizes only the high molecular MAP2 forms, MAP2A and MAP2B. For long term storage, store at 4°C.

MAP2 (Microtubule Associated Protein 2) exists in two high molecular weight forms (MAP2a & MAP2b) and a low molecular weight form. The expression of MAP2 is developmentally regulated and its multiple forms arise by alternative splicing of a single gene. MAP2 is involved in microtubule assembly, which is an essential step in neurogenesis. The products of similar genes in rat and mouse are neuron-specific cytoskeletal proteins that are enriched in dendrites, implicating a role in determining and stabilizing dendritic shape during neuron development. MAP2 is useful in studies of neuron structure in normal and malignant brain tissue as well as in degenerative diseases such as Alzheimer's Disease (AD).
TRUSTED_SUSTAINABILITY

Specifications

Antigen MAP2
Applications Immunohistochemistry, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene Map2
Gene Accession No. P11137, P15146, P20357
Gene Alias a730034c02; AC091465.1; DKFZp686I2148; G1-397-34; MAP 2; Map2; MAP-2; map2.S; map2a; map-2a; map2ab; MAP2B; MAP2C; MAP2R; microtubule associated protein 2; microtubule associated protein 2 S homeolog; microtubule associated protein 2; microtubule-associated protein 2; microtubule associated protein 2C; microtubule-associated protein 2; microtubule-associated protein-2; Mtap2; Mtap-2; OTTHUMP00000163916; OTTHUMP00000208143; repro4; XELAEV_18047318mg; xmap2
Gene Symbols Map2
Host Species Chicken
Immunogen Mix of recombinant human constructs of projection domain sequences, amino acids 235-1588.
Purification Method PEG precipitation
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 17756, 25595, 4133
Target Species Human, Mouse, Rat
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgY
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.