Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MAP2K1, Rabbit anti-Human, FITC, Clone: H8, Abnova™

Catalog No. 89335774 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 Reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-335-774 10 Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-335-774 Supplier Abnova Corporation Supplier No. MAB18940
Only null left
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Specifications

Antigen MAP2K1
Applications Flow Cytometry
Classification Monoclonal
Clone H8
Conjugate FITC
Dilution Flow Cytometry (5 uL/106 cells) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (0.2% BSA, 0.09% sodium azide).
Gene mitogen-activated protein kinase kinase 1
Gene Alias MAPKK1/MEK1/MKK1/PRKMK1
Gene Symbols MAP2K1
Host Species Rabbit
Immunogen A synthetic phosphopeptide corresponding to residues surrounding S298 of human MAP2K1.
Quantity 10 Reactions
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5604
Target Species Human, Rat
Content And Storage Store at 4°C.
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.