Learn More
MAPA™ KryTech 395 Heavy Duty Work Gloves
Catalog No. 19131340 Shop All MAPA Professional Products
Change view
Click to view available options
Size:
7
8
9
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-131-340 7
19-131-342 9
19-131-341 8
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 19-131-340 Supplier MAPA™ Supplier No. 395317
This item is not returnable. View return policy
Multi-purpose gloves protect users against cuts, liquids, and contact heat up to 100°C. Nitrile coating enhances grip capabilities in wet or slippery applications.
  • Combined protection against cuts, liquid, and contact heat up to 100°C
  • Nitrile coating enhances grip capabilities in wet or slippery applications
  • Entirely liquid-proof, withstanding mechanical aggression
  • Micro-organism protection against bacteria and fungi
  • Silicone-free to prevent traces and defects on metal sheets or glass
  • Proven long-lasting performance
Specifications
Size 7