Learn More
MAPA™ SensiTron™ Sheer Latex Gloves
Catalog No. 19048698E Shop All MAPA Professional Products
Change view
Click to view available options
Size:
7
7.5
8
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-048-698E 8
19-048-698C 7
19-048-698D 7.5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 19-048-698E Supplier MAPA™ Supplier No. 516565
This item is not returnable. View return policy
Sensitron™ manufacturing cleaning process ensures consistent low level particles, extractables and non-volatile residues for Class 100/ISO 5 and higher controlled environments.
Specifications
Cleanroom Class Class 100 (ISO 5)
Color Natural
Disposable Single-use
Exterior Finish Fully Textured
Finger Thickness (English) 8 mil
Finger Thickness (Metric) 0.20 mm
Hand Specific
Material Latex
Size 8
Sterility Sterile
Thickness (English) 8 mil
Length (English) 12 in.
Length (Metric) 30.5 cm