Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MAPA™ Solo Ultra 999 Nitrile Gloves

Catalog No. 191302651E Shop All MAPA Professional Products
Change view
Click to view available options
Size:
Medium
X-Large
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-130-2651E X-Large
19-130-2651C Medium
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-130-2651E Supplier MAPA™ Supplier No. 999419
This item is not returnable. View return policy

White, disposable Solo 999 Nitrile Gloves are ideal for environments where handling oily or precision parts is necessary.

  • Textured fingertips for improved dexterity
  • Easy to don on and take off
  • Latex-free
  • Previous name was Solo Ultra 999
  • Ideal for automotive/mechanics industry, aeronautics industry and laboratories

Specifications

Certifications/Compliance FDA 21 CFR 177.2600
Chemotherapeutic Rated No
Applications Laboratory, Industrial
Color White
Disposable Single-use
Exterior Finish Partially Textured
Laboratory/Cleanroom Use No
Latex Free Yes
Material Nitrile
Powdered Powder Free
Size X-Large
Thickness (English) 4 mil
Thickness (Metric) 0.10 mm
Model 99419
Series Solo 999
Length (English) 12 in.
Length (Metric) 30.5 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.