Learn More
Description
Unsupported powder-free fluoroelastomer/nitrile gloves resist aromatic solvents, chloroform, and PCBs. Suitable for use in refineries, plating operations, laboratories, and hazardous material handling.
- Consist of a layer of black FluoroSolv fluoroelastomer polymer over a green, powder-free nitrile shell
- Two-tone layering serves as a visual wear indicator
- Nitrile layer also provides increased cut, puncture, and tear resistance
- Z-pattern texture channels chemicals away from glove surface, improving wet and dry grip
- Hand-specific and unlined with a flat cuff
- Embossed with size measurement for easy identification
- Style F-466 is 11 in. long (28 cm), 15 mil thick
- Style F-467 is 14 in. long (36 cm), 25 mil thick
Not for use with ketonic solvents.
Specifications
Specifications
| Powdered | Powder Free |
| Size | 11 |
| Length (English) | 11 in. |
| Length (Metric) | 28 cm |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.