Learn More
MAPA™ FluoTech 344 Fluoroelastomer Gloves, Cotton Knit Liner
Catalog No. 19165708 Shop All MAPA Professional Products
Change view
Click to view available options
:
Black; 8
Catalog No.
19-165-708 Black; 8
1 options
This item is not returnable. View return policy
Catalog No. 19-165-708 Supplier MAPA™ Supplier No. 344318
This item is not returnable. View return policy
Fluoroelastomer gloves with cotton knit liner are comfortable and flexible for extended wear.
  • Cotton-knit interior
  • High mechanical resistance
  • Long service life due to textile support
  • Site pollution control
  • Printing
  • Petrochemical refining
  • Emergency interventions on chemical accidents
Specifications
Certifications/Compliance (Europe) Cat 3 CE 0334, EN 420
Color Black
Latex Free Yes
Material Fluoroelastomer
Size 8
EN388 Standard EN 388:3121X
EN ISO 374-1 Type A
EN ISO 374-1 CHEMICAL LETTER CODES ACDEFGJLMN
EN ISO 374 - 5 Bacteria, Fungi only
Glove Cuff Style Pinked
Grip Design Smooth
Liner Material Cotton Knit
Lining Lined
Model 344318
Series FluoTech 344
Length (English) 14 in.
Length (Metric) 35.5 cm