Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MAPA™ Ultrane 551 Lightweight Polyurethane Coated Seamless Knit Gloves

Catalog No. 19161407 Shop All MAPA Professional Products
Change view
Click to view available options
Size:
6
7
8
9
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-161-407 9
19-161-404 6
19-161-405 7
19-161-406 8
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 19-161-407 Supplier MAPA™ Supplier No. 551409
This item is not returnable. View return policy

Seamless knit lining absorbs perspiration for increased user comfort, while the polyurethane coating enhances grip capabilities.

  • Reduced glove thickness increases dexterity, while still providing abrasion resistance
  • Seamless knit lining absorbs perspiration for increased user comfort
  • Silicone free to prevent traces, defects on metal sheets and glass prior to painting
  • Contains traces of DMF in compliance with occupational exposure limits
  • Free of harmful substances: STANDARD 100 by OEKO-TEX™

Specifications

Construction Seamless Knit
Coating Material Polyurethane Foam
Color Gray
Exterior Finish No Texture
Hand Specific
Product Type Seamless Knit Gloves
Size 9
EN388 Standard 4131X
For Use With (Application) Automotive Assembly, Electronics Repair, and Construction
Series Ultrane 551
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.