Learn More
MAPK13 Mouse anti-Human, Clone: OTI2A10, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF505856

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
The protein encoded by this gene is a member of the MAP kinase family. MAP kinases act as an integration point for multiple biochemical signals, and are involved in a wide variety of cellular processes such as proliferation, differentiation, transcription regulation and development. This kinase is closely related to p38 MAP kinase, both of which can be activated by proinflammatory cytokines and cellular stress. MAP kinase kinases 3, and 6 can phosphorylate and activate this kinase. Transcription factor ATF2, and microtubule dynamics regulator stathmin have been shown to be the substrates of this kinase.Specifications
MAPK13 | |
Monoclonal | |
Unconjugated | |
MAPK13 | |
MAPK 13, MAPK-13, PRKM13, SAPK4, p38delta | |
Mouse | |
Affinity Chromatography | |
RUO | |
5603 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
OTI2A10 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
O15264 | |
MAPK13 | |
Full length human recombit protein of human MAPK13 produced in HEK293T cell. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2b |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.