Learn More
Invitrogen™ MAPKAP1 Polyclonal Antibody

Description
Suggested positive control: human skeletal muscle and heart protein or cell lysate. Immunogen sequence: NTAQRLERLR KERQNQIK.
Specifications
Specifications
| Antigen | MAPKAP1 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.2% gelatin and 0.05% sodium azide |
| Gene | MAPKAP1 |
| Gene Accession No. | A2VDU2, Q6AYF1, Q8BKH7, Q9BPZ7, Q9W6S3 |
| Gene Alias | AI591529; D230039K05Rik; im:7143088; JC310; MAPK associated protein 1; mapkap1; MEKK2-interacting protein 1; MGC2745; Mip1; MIP1SIN1SIN1b; mitogen activated protein kinase associated protein 1; mitogen-activated protein kinase 2-associated protein 1; mitogen-activated protein kinase associated protein 1; mSIN1; ras inhibitor MGC2745; RP11-269P11.1; SAPK interacting protein; SAPK-interacting protein 1; si:ch73-242f23.1; SIN1; SIN1b; SIN1g; stress-activated map kinase interacting protein 1; stress-activated map kinase-interacting protein 1; stress-activated protein kinase-interacting 1; target of rapamycin complex 2 subunit MAPKAP1; TORC2 subunit MAPKAP1 |
| Gene Symbols | MAPKAP1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.