Learn More
Selleck Chemical LLC Maraviroc 100mg 376348-65-1 UK-427857
Supplier: Selleck Chemical LLC S2003100MG
Maraviroc (UK-427857) is a CCR5 antagonist for MIP-1?, MIP-1? and RANTES with IC50 of 3.3 nM, 7.2 nM and 5.2 nM in cell-free assays, respectively. Maraviroc is used in the treatment of HIV infection. *For Research & Development use only. Product is not intended for drug, household, or human consumption.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.