Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MARCH1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA553199
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA553199 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA553199 Supplier Invitrogen™ Supplier No. PA553199
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KVYVQLWRRL KAYNRVIFVQ NCPDTAKKLE KNFSCNVNTD IKDAVVVPVP QTGANSLPSA EGG Highest antigen sequence identity to the following orthologs: Mouse - 89%, Rat - 87%.

E3 ubiquitin-protein ligase that mediates ubiquitination of TFRC, CD86, FAS and MHC class II proteins, such as HLA-DR alpha and beta, and promotes their subsequent endocytosis and sorting to lysosomes via multivesicular bodies. By constitutively ubiquitinating MHC class II proteins in immature dendritic cells, down-regulates their cell surface localization thus sequestering them in the intracellular endosomal system.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MARCH1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MARCH1
Gene Accession No. Q8TCQ1
Gene Alias 1-Mar; 2900024D24Rik; BB085186; E3 ubiquitin-protein ligase MARCH1; MARCH1; MARCHF1; MARCH-I; membrane associated ring finger 1; membrane associated ring-CH-type finger 1; membrane-associated ring finger (C3HC4) 1; membrane-associated ring finger (C3HC4) 1, E3 ubiquitin protein ligase; membrane-associated RING finger protein 1; Membrane-associated RING-CH protein I; RGD1305148; RING finger protein 171; RING-type E3 ubiquitin transferase MARCH1; RNF171
Gene Symbols MARCH1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human MARCH1. Recombinant protein control fragment (Product #RP-90986).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55016
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.