Learn More
Invitrogen™ MARCH2 Polyclonal Antibody
Description
Recommended positive controls: HepG2 membrane. Predicted reactivity: Mouse (94%), Rat (94%), Zebrafish (81%), Xenopus laevis (80%), Pig (95%), Bovine (91%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Specifications
Specifications
| Antigen | MARCH2 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 |
| Gene | MARCHF2 |
| Gene Accession No. | Q9P0N8 |
| Gene Alias | 9530046H09Rik; E3 ubiquitin-protein ligase MARCH2; HSPC240; MARCH2; MARCHF2; MARCH-II; membrane associated ring finger 2; membrane associated RING-CH finger protein II; membrane associated ring-CH-type finger 2; membrane-associated ring finger (C3HC4) 2; membrane-associated ring finger (C3HC4) 2, E3 ubiquitin protein ligase; membrane-associated RING finger protein 2; Membrane-associated RING-CH protein II; RGD1306395; RING finger protein 172; RING-type E3 ubiquitin transferase MARCH2; RNF172; syntaxin-6-binding protein |
| Gene Symbols | MARCHF2 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.