Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MARCH8 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA542272
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA542272 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA542272 Supplier Invitrogen™ Supplier No. PA542272
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: MSMPLHQISA IPSQDAISAR VYRSKTKEKE REEQNEKTLG HFMSHSSNIS Sequence homology: Cow: 93%; Dog: 79%; Human: 100%; Mouse: 100%; Rat: 100%.

MARCH8 (c-MIR) is a novel E3 ubiquitin ligase designated as the modulator of immune recognition (MIR) family, whose catalytic domain is a variant RING domain (RING-CH domain). MARCH8 was found as a functional and structural homolog of KSHV MIR1 and MIR2. MARCH8 targets B7-2 to lysosomal degradation and down-regulates the B7-2 surface expression through ubiquitination of its cytoplasmic tail. Furthermore, MARCH8 has been shown to down-regulate the expression of transferrin receptor and Fas, an important molecule for the induction of apoptosis. MARCH8 is the first example of an E3 ubiquitin ligase that is capable of inhibiting MHC II expression. Recent findings suggest that MARCH8 may regulate immune responses by promoting ubiquitination of MHC-II and CD86, leading to their subsequent endocytosis and lysosomal degradation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MARCH8
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene MARCH8
Gene Accession No. Q5T0T0
Gene Alias 1300017E09Rik; cellular modulator of immune recognition; cellular modulator of immune recognition (c-MIR); CMIR; c-MIR; c-mir, cellular modulator of immune recognition; E3 ubiquitin-protein ligase MARCH8; E3 ubiquitin-protein ligase MARCHF8; E3 ubiquitin-protein ligase MARCH8; March8; MARCHF8; MARCH-VIII; membrane associated ring finger 8; membrane associated ring-CH-type finger 8; membrane-associated ring finger (C3HC4) 8; membrane-associated ring finger (C3HC4) 8, E3 ubiquitin protein ligase; membrane-associated RING finger protein 8; Membrane-associated RING-CH protein VIII; MIR; RING finger protein 178; RING-type E3 ubiquitin transferase MARCH8; RING-type E3 ubiquitin transferase MARCHF8; RNF178; RP11-67C2.1
Gene Symbols MARCH8
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human MARCH8 (aa 1-50).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 220972
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.